Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004836.1 | NP_001004836.1 | 3 - 4 | 326 | 90 | |
GenBank mRNA | BC074636.1 | - | 3 - 4 | 326 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |