Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mad2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244840.1 | NP_001231769.1 | 8 - 9 | 634 | 77 | |
NM_001244841.1 | NP_001231770.1 | 7 - 8 | 570 | 77 | ||
NM_001244842.1 | NP_001231771.1 | 7 - 8 | 594 | 77 | ||
NR_045201.1 | 7 - 8 | 563 | 77 | |||
XM_012966356.2 | XP_012821810.1 | 8 - 9 | 661 | 77 | ||
XM_018095335.1 | XP_017950824.1 | 8 - 9 | 1085 | 77 | ||
XM_018095336.1 | XP_017950825.1 | 8 - 9 | 645 | 77 | ||
XM_018095337.1 | XP_017950826.1 | 7 - 8 | 584 | 77 | ||
GenBank mRNA | BC074654.1 | - | 7 - 8 | 552 | 77 | |
BQ521961.1 | - | 7 - 8 | 552 | 77 | ||
CR760993.2 | - | 7 - 8 | 582 | 77 | ||
CX840685.1 | - | 7 - 8 | 593 | 77 | ||
CX842973.1 | - | 6 - 7 | 519 | 77 | ||
ES678430.1 | - | 8 - 9 | 634 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |