Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sfrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004846.1 | NP_001004846.1 | 1 - 2 | 818 | 92 | |
GenBank mRNA | BC074661.1 | - | 1 - 2 | 818 | 92 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |