Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nfkbia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004851.1 | NP_001004851.1 | 5 - 6 | 1027 | 102 | |
GenBank mRNA | BC074672.1 | - | 5 - 6 | 1027 | 102 | |
CR760347.2 | - | 5 - 6 | 1016 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |