Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004854.1 | NP_001004854.1 | 6 - 7 | 783 | 97 | |
XM_012960808.2 | XP_012816262.1 | 6 - 7 | 595 | 97 | ||
GenBank mRNA | BC074675.1 | - | 6 - 7 | 783 | 97 | |
CR761982.2 | - | 6 - 7 | 783 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |