Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004855.1 | NP_001004855.1 | 1 - 2 | 192 | 111 | |
XM_012962865.1 | XP_012818319.1 | 2 - 3 | 362 | 111 | ||
XM_012962866.2 | XP_012818320.1 | 2 - 3 | 293 | 111 | ||
XM_012962867.1 | XP_012818321.1 | 2 - 3 | 218 | 111 | ||
XM_012962868.2 | XP_012818322.1 | 2 - 3 | 211 | 111 | ||
XM_012962869.2 | XP_012818323.1 | 2 - 3 | 207 | 111 | ||
GenBank mRNA | BC074676.1 | - | 1 - 2 | 192 | 111 | |
CR760910.2 | - | 1 - 2 | 172 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |