Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004858.1 | NP_001004858.1 | 5 - 6 | 717 | 102 | |
XM_012959055.2 | XP_012814509.1 | 5 - 6 | 848 | 102 | ||
GenBank mRNA | BC074682.1 | - | 5 - 6 | 717 | 102 | |
CT010555.2 | - | 5 - 6 | 705 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |