Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004859.1 | NP_001004859.1 | 6 - 7 | 551 | 90 | |
XM_012968198.2 | XP_012823652.1 | 6 - 7 | 616 | 90 | ||
GenBank mRNA | BC074687.1 | - | 6 - 7 | 551 | 90 | |
CR855591.2 | - | 6 - 7 | 529 | 90 | ||
CR926234.1 | - | 6 - 7 | 533 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |