Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: n4bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004870.1 | NP_001004870.1 | 2 - 3 | 1638 | 95 | |
XM_012960812.2 | XP_012816266.1 | 2 - 3 | 1788 | 95 | ||
GenBank mRNA | BC075104.1 | - | 2 - 3 | 1638 | 95 | |
CT025272.2 | - | 2 - 3 | 1614 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |