Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004885.2 | NP_001004885.1 | 6 - 7 | 663 | 86 | |
XM_012959343.2 | XP_012814797.1 | 7 - 8 | 769 | 86 | ||
XM_012959347.2 | XP_012814801.1 | 7 - 8 | 766 | 86 | ||
GenBank mRNA | BC075282.1 | - | 6 - 7 | 665 | 86 | |
CR855614.2 | - | 6 - 7 | 662 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |