Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chchd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004897.1 | NP_001004897.1 | 3 - 4 | 398 | 96 | |
XM_012958817.2 | XP_012814271.1 | 2 - 3 | 308 | 96 | ||
XM_012958818.2 | XP_012814272.1 | 2 - 3 | 308 | 96 | ||
XM_012958819.2 | XP_012814273.1 | 2 - 3 | 208 | 96 | ||
GenBank mRNA | BC075310.1 | - | 3 - 4 | 398 | 96 | |
CR760954.2 | - | 2 - 3 | 288 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |