Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chchd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004897.1 | NP_001004897.1 | 4 - 5 | 482 | 81 | |
XM_012958817.2 | XP_012814271.1 | 3 - 4 | 392 | 81 | ||
XM_012958818.2 | XP_012814272.1 | 3 - 4 | 392 | 81 | ||
XM_012958819.2 | XP_012814273.1 | 3 - 4 | 292 | 81 | ||
GenBank mRNA | BC075310.1 | - | 4 - 5 | 482 | 81 | |
CR760954.2 | - | 3 - 4 | 372 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |