Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004904.1 | NP_001004904.2 | 1751 | 92 | ||
GenBank mRNA | BC075328.1 | - | 1751 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis |