Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdzd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004906.1 | NP_001004906.1 | 2 - 3 | 228 | 71 | |
GenBank mRNA | BC075334.1 | - | 2 - 3 | 228 | 71 | |
CR760601.2 | - | 2 - 3 | 236 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |