Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdzd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004906.1 | NP_001004906.1 | 4 - 5 | 378 | 88 | |
XM_018096199.1 | XP_017951688.1 | 3 - 4 | 358 | 88 | ||
GenBank mRNA | BC075334.1 | - | 4 - 5 | 378 | 88 | |
CR760601.2 | - | 4 - 5 | 386 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |