Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bicdl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004934.1 | NP_001004934.1 | 1005 | 66 | ||
GenBank mRNA | BC075409.1 | - | 1005 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |