Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004949.2 | NP_001004949.1 | 7 - 8 | 692 | 94 | |
XM_012966386.2 | XP_012821840.1 | 8 - 9 | 906 | 94 | ||
GenBank mRNA | BC075438.1 | - | 7 - 8 | 693 | 94 | |
CR760856.2 | - | 7 - 8 | 708 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |