Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gata3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004967.1 | NP_001004967.1 | 4 - 5 | 1471 | 79 | |
XM_012958832.2 | XP_012814286.1 | 4 - 5 | 1595 | 79 | ||
XM_012958833.2 | XP_012814287.1 | 4 - 5 | 1592 | 79 | ||
XM_012958834.2 | XP_012814288.1 | 4 - 5 | 1591 | 79 | ||
XM_012958835.2 | XP_012814289.1 | 4 - 5 | 1530 | 79 | ||
GenBank mRNA | BC075479.1 | - | 4 - 5 | 1471 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |