Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004981.1 | NP_001004981.1 | 3 - 4 | 425 | 76 | |
XM_018092030.1 | XP_017947519.1 | 2 - 3 | 390 | 76 | ||
GenBank mRNA | BC075530.1 | - | 3 - 4 | 425 | 76 | |
CR762186.2 | - | 3 - 4 | 373 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |