Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yeats4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004981.1 | NP_001004981.1 | 5 - 6 | 602 | 100 | |
XM_018092030.1 | XP_017947519.1 | 4 - 5 | 567 | 100 | ||
GenBank mRNA | BC075530.1 | - | 5 - 6 | 602 | 100 | |
CR762186.2 | - | 5 - 6 | 550 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |