Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ints13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004984.1 | NP_001004984.1 | 1416 | 96 | ||
GenBank mRNA | BC075538.1 | - | 1416 | 96 | ||
CR761707.2 | - | 1343 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis |