Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005002.1 | NP_001005002.1 | 2 - 3 | 557 | 59 | |
GenBank mRNA | BC076648.1 | - | 2 - 3 | 557 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |