Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005015.1 | NP_001005015.1 | 10 - 11 | 1378 | 93 | |
GenBank mRNA | BC076682.1 | - | 10 - 11 | 1378 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |