Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005023.1 | NP_001005023.1 | 2 - 3 | 166 | 61 | |
XM_012960866.2 | XP_012816320.1 | 2 - 3 | 202 | 61 | ||
GenBank mRNA | BC076703.1 | - | 2 - 3 | 166 | 61 | |
CT027947.2 | - | 2 - 3 | 144 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |