Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005028.1 | NP_001005028.1 | 4 - 5 | 692 | 68 | |
XM_012966417.2 | XP_012821871.1 | 5 - 6 | 769 | 68 | ||
XM_012966418.1 | XP_012821872.1 | 5 - 6 | 769 | 68 | ||
XM_012966421.2 | XP_012821875.1 | 4 - 5 | 789 | 68 | ||
GenBank mRNA | BC076711.1 | - | 4 - 5 | 692 | 68 | |
CT010525.2 | - | 4 - 5 | 498 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |