Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: isoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005029.1 | NP_001005029.1 | 2 - 3 | 466 | 97 | |
XM_012966424.2 | XP_012821878.1 | 2 - 3 | 504 | 97 | ||
XM_012966425.2 | XP_012821879.1 | 2 - 3 | 505 | 97 | ||
XM_018095350.1 | XP_017950839.1 | 3 - 4 | 603 | 97 | ||
GenBank mRNA | BC076875.1 | - | 2 - 3 | 466 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |