Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005055.2 | NP_001005055.2 | 4 - 5 | 799 | 69 | |
GenBank mRNA | BC076943.1 | - | 4 - 5 | 799 | 69 | |
CU075748.1 | - | 4 - 5 | 460 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |