Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: traf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005074.1 | NP_001005074.1 | 4 - 5 | 693 | 88 | |
XM_018091173.1 | XP_017946662.1 | 4 - 5 | 2342 | 88 | ||
GenBank mRNA | BC076992.1 | - | 4 - 5 | 693 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |