Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005082.1 | NP_001005082.1 | 5 - 6 | 416 | 86 | |
XM_012962897.2 | XP_012818351.1 | 5 - 6 | 490 | 86 | ||
GenBank mRNA | BC077004.1 | - | 5 - 6 | 416 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |