Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pomp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005093.2 | NP_001005093.1 | 5 - 6 | 460 | 64 | |
GenBank mRNA | BC077023.1 | - | 5 - 6 | 462 | 64 | |
CR855731.2 | - | 5 - 6 | 410 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |