Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011000.1 | NP_001011000.1 | 2 - 3 | 224 | 92 | |
XM_018093865.1 | XP_017949354.1 | 2 - 3 | 212 | 92 | ||
GenBank mRNA | BC082500.1 | - | 2 - 3 | 224 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |