Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpy30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011017.1 | NP_001011017.1 | 4 - 5 | 414 | 83 | |
GenBank mRNA | BC082731.1 | - | 4 - 5 | 414 | 83 | |
CR760705.1 | - | 3 - 4 | 259 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |