Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011024.1 | NP_001011024.1 | 4 - 5 | 426 | 71 | |
GenBank mRNA | BC082740.1 | - | 4 - 5 | 426 | 71 | |
CR761928.2 | - | 4 - 5 | 424 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |