Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011032.1 | NP_001011032.1 | 4 - 5 | 743 | 76 | |
GenBank mRNA | AY714337.1 | - | 4 - 5 | 743 | 76 | |
BC171265.1 | - | 4 - 5 | 718 | 76 | ||
CT027924.3 | - | 4 - 5 | 741 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |