Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc30a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011041.1 | NP_001011041.1 | 1 - 2 | 198 | 86 | |
XM_018094625.1 | XP_017950114.1 | 1 - 2 | 354 | 86 | ||
GenBank mRNA | BC084148.1 | - | 1 - 2 | 198 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |