Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cebpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011044.1 | NP_001011044.1 | 1 | 954 | 67 | |
GenBank mRNA | BC084168.1 | - | 1 | 954 | 67 | |
BC167260.1 | - | 1 | 957 | 67 | ||
CT030439.1 | - | 1 | 963 | 67 | ||
FJ771049.1 | - | 1 | 963 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Both primers and probe map within a single exon |