Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pacsin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011050.1 | NP_001011050.1 | 6 - 7 | 895 | 67 | |
GenBank mRNA | BC084175.1 | - | 6 - 7 | 895 | 67 | |
CT030501.1 | - | 6 - 7 | 898 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |