Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pacsin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011050.1 | NP_001011050.1 | 8 - 9 | 1150 | 77 | |
GenBank mRNA | BC084175.1 | - | 8 - 9 | 1150 | 77 | |
CT030501.1 | - | 8 - 9 | 1153 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |