Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plscr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011298.1 | NP_001011298.1 | 422 | 87 | ||
GenBank mRNA | BC088016.1 | - | 422 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |