Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203529.1 | NP_988860.1 | 2 - 3 | 343 | 73 | |
GenBank mRNA | BC088005.1 | - | 2 - 3 | 325 | 73 | |
M32454.1 | - | 2 - 3 | 343 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |