Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl7l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203555.1 | NP_988886.1 | 809 | 67 | ||
GenBank mRNA | BC059773.1 | - | 809 | 67 | ||
CR760519.2 | - | 748 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis |