Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v0c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203562.1 | NP_988893.1 | 2 - 3 | 407 | 77 | |
GenBank mRNA | BC059745.1 | - | 2 - 3 | 407 | 77 | |
CT030632.1 | - | 2 - 3 | 270 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |