Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tm4sf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203627.1 | NP_988958.1 | 3 - 4 | 474 | 134 | |
GenBank mRNA | BC061278.1 | - | 3 - 4 | 474 | 134 | |
CR760805.1 | - | 3 - 4 | 432 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |