Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203642.1 | NP_988973.1 | 6 - 7 | 821 | 67 | |
GenBank mRNA | BC061386.1 | - | 6 - 7 | 821 | 67 | |
CR848567.2 | - | 6 - 7 | 613 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |