Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203645.1 | NP_988976.1 | 6 - 7 | 614 | 73 | |
XM_012953192.2 | XP_012808646.1 | 6 - 7 | 674 | 73 | ||
GenBank mRNA | BC061371.1 | - | 6 - 7 | 614 | 73 | |
CR761448.2 | - | 5 - 6 | 468 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |