Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203655.1 | NP_988986.1 | 5 - 6 | 646 | 78 | |
GenBank mRNA | BC061319.1 | - | 5 - 6 | 646 | 78 | |
CR760865.2 | - | 5 - 6 | 644 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |