Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ykt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203692.1 | NP_989023.1 | 2 - 3 | 260 | 79 | |
XM_012958755.2 | XP_012814209.1 | 2 - 3 | 242 | 79 | ||
GenBank mRNA | BC061415.1 | - | 2 - 3 | 260 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |