Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnot9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203693.1 | NP_989024.1 | 4 - 5 | 699 | 88 | |
GenBank mRNA | BC061412.1 | - | 4 - 5 | 699 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |