Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnot9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203693.1 | NP_989024.1 | 7 - 8 | 992 | 94 | |
GenBank mRNA | BC061412.1 | - | 7 - 8 | 992 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |