Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203710.1 | NP_989041.1 | 2 - 3 | 333 | 95 | |
XM_012968151.2 | XP_012823605.1 | 1 - 2 | 470 | 95 | ||
XM_012968152.2 | XP_012823606.1 | 1 - 2 | 473 | 95 | ||
GenBank mRNA | BC061372.1 | - | 2 - 3 | 333 | 95 | |
CR762001.2 | - | 1 - 2 | 127 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |